Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-4-Fluorophenylglycine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
500mg
95%
C23H18FNO4
MFCD01320845
(S)-N-FMOC-4-Fluorophenylglycine, 95%, (98% e.e.)
95%
C23H18FNO4
MFCD01320845
(S)-N-FMOC-4-Fluorophenylglycine, 95%, (98% e.e.)
Color White
Quantity 500mg
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
CAS 678988-18-6
Synonym fmoc-4-fluoro-l-phenylglycine,n-fmoc-4-fluorophenylglycine,2s-2-9h-fluoren-9-ylmethoxycarbonylamino-2-4-fluorophenyl acetic acid,s-n-fmoc-4-fluorophenylglycine,s-9h-fluoren-9-ylmethoxy carbonyl amino 4-fluorophenyl acetic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-4-fluorophenyl acetic acid,s-9h-fluoren-9-ylmethoxycarbonylamino-4-fluoro-phenyl-acetic acid,s-n-9-fluorenylmethoxycarbonyl-alpha-4-fluorophenyl acetic acid
InChI Key UUEGPRZUDVECNF-UHFFFAOYNA-N
Molecular Weight (g/mol) 391.40
PubChem CID 68968394
Formula Weight 391.39
Physical Form Powder
Percent Purity ≥94%
Show More Show Less