Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass Bottle
136552-06-2
C19H17NO4
323.35
MFCD01320843
BXRZCDISGRVJCA-KRWDZBQOSA-N
1-fmoc-s-azetidine-2-carboxylic acid,s-1-9h-fluoren-9-yl methoxy carbonyl azetidine-2-carboxylic acid,s-n-fmoc-azetidine-2-carboxylic acid,fmoc-aze-oh,fmoc-s-azetidine-2-carboxylic acid,2s-1-9h-fluoren-9-ylmethoxycarbonyl azetidine-2-carboxylic acid,2s-1-9h-fluoren-9-ylmethoxy carbonyl azetidine-2-carboxylic acid,2r-1-9h-fluoren-9-ylmethoxycarbonyl azetidine-2-carboxylate,fmoc-l-azetidine-2-carboxylic acid,s-n-fmoc-azetidine-2-carboxylicacid
2734468
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)azetidine-2-carboxylic acid
C1CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
136552-06-2
C19H17NO4
323.35
MFCD01320843
BXRZCDISGRVJCA-KRWDZBQOSA-N
1-fmoc-s-azetidine-2-carboxylic acid,s-1-9h-fluoren-9-yl methoxy carbonyl azetidine-2-carboxylic acid,s-n-fmoc-azetidine-2-carboxylic acid,fmoc-aze-oh,fmoc-s-azetidine-2-carboxylic acid,2s-1-9h-fluoren-9-ylmethoxycarbonyl azetidine-2-carboxylic acid,2s-1-9h-fluoren-9-ylmethoxy carbonyl azetidine-2-carboxylic acid,2r-1-9h-fluoren-9-ylmethoxycarbonyl azetidine-2-carboxylate,fmoc-l-azetidine-2-carboxylic acid,s-n-fmoc-azetidine-2-carboxylicacid
2734468
(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)azetidine-2-carboxylic acid
C1CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Melting Point 135°C
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 500mg
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 323.35
Physical Form Powder
Percent Purity 95%,98% ee
Chemical Name or Material (S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, (98% e.e.)
Show More Show Less