Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
169566-81-8,1634-04-4
C21H23NO4
353.42
AWEZXIRZNQCCNN-NRFANRHFSA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid,fmoc-me-val-oh,fmoc-a-me-val-oh,fmoc-s-alpha-methylvaline,s-2-fmoc-amino-2,3-dimethylbutanoic acid,l-valine,n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl,n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-d-isovaline,2s-2-9h-fluoren-9-ylmethoxycarbonylamino-2,3-dimethylbutanoic acid,s-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid
11394064
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
169566-81-8,1634-04-4
C21H23NO4
353.42
AWEZXIRZNQCCNN-NRFANRHFSA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid,fmoc-me-val-oh,fmoc-a-me-val-oh,fmoc-s-alpha-methylvaline,s-2-fmoc-amino-2,3-dimethylbutanoic acid,l-valine,n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl,n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-d-isovaline,2s-2-9h-fluoren-9-ylmethoxycarbonylamino-2,3-dimethylbutanoic acid,s-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid
11394064
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Melting Point 37°C to 53°C
Specific Rotation 8.4
Infrared Spectrum Authentic
Enantiomeric Excess 98.0% min.
Color White to Yellow
Residual Solvents 10.0% max. (MTBE)
Quantity 500mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 353.42
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (S)-N-FMOC-α-Methylvaline, 98% ee
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning