Alfa Aesar™ S-n-Propyl thioacetate, 98+%

Manufacturer: Alfa Aesar™ B2187514

Quantity 25g

 View more versions of this product

Catalog No. AAB2187514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2307-10-0
    Molecular Formula C5H10OS
    Formula Weight 118.2
    MDL Number MFCD00039937
    UN Number UN3272
    Beilstein 1740765
    Synonym Thioacetic acid S-n-propyl ester
    Chemical Name or Material S-n-Propyl thioacetate

    Specifications

    Formula Weight 118.2
    MDL Number MFCD00039937
    CAS 2307-10-0
    UN Number UN3272
    Beilstein 1740765
    Synonym Thioacetic acid S-n-propyl ester
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 137° to 139°C
    Quantity 25g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Molecular Formula C5H10OS
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Chemical Name or Material S-n-Propyl thioacetate
    Assay ≥98%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit