Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ S-n-Propyl thioacetate, 98+%

Click to view available options
Quantity:
5g
25g
100g
2307-10-0
C5H10OS
118.194
MFCD00039937
SBWFWBJCYMBZEY-UHFFFAOYSA-N
s-propyl thioacetate,propyl thioacetate,ethanethioic acid, s-propyl ester,propyl thiolacetate,s-n-propyl thioacetate,acetic acid, thio-, s-propyl ester,unii-y7284nz6ep,1-propylsulfanyl ethanone,n-propyl thio acetate,fema no. 3385
61295
S-propyl ethanethioate
CCCSC(=O)C
2307-10-0
C5H10OS
118.194
MFCD00039937
SBWFWBJCYMBZEY-UHFFFAOYSA-N
s-propyl thioacetate,propyl thioacetate,ethanethioic acid, s-propyl ester,propyl thiolacetate,s-n-propyl thioacetate,acetic acid, thio-, s-propyl ester,unii-y7284nz6ep,1-propylsulfanyl ethanone,n-propyl thio acetate,fema no. 3385
61295
S-propyl ethanethioate
CCCSC(=O)C
Boiling Point 137°C to 139°C
Quantity 5g
UN Number UN3272
Beilstein 1740765
Formula Weight 118.2
Percent Purity ≥98%
Chemical Name or Material S-n-Propyl thioacetate

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.