Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Neomenthyldiphenylphosphine, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
43077-29-8
C22H29P
324.448
MFCD00013415
BEYDOEXXFGNVRZ-COPCDDAFSA-N
s-+-neomenthyldiphenylphosphine,1s,2s,5r-2-isopropyl-5-methylcyclohexyl diphenylphosphine,s-nmdpp,+-neomenthyldiphenylphosphine,1s,2s,5r-2-isopropyl-5-methylcyclohexyl-diphenylphosphine,1s,2s,5r-2-isopropyl-5-methylcyclohexyl diphenylphosphane,+-nmdpp,1r,3s,4s-p-menthane-3-yl diphenylphosphine,1s,2s,5r-5-methyl-2-propan-2-ylcyclohexyl-diphenylphosphane,1s,2s,5r-5-methyl-2-propan-2-yl cyclohexyl diphenyl phosphane
2734569
[(1S,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl]-diphenylphosphane
CC1CCC(C(C1)P(C2=CC=CC=C2)C3=CC=CC=C3)C(C)C
43077-29-8
C22H29P
324.448
MFCD00013415
BEYDOEXXFGNVRZ-COPCDDAFSA-N
s-+-neomenthyldiphenylphosphine,1s,2s,5r-2-isopropyl-5-methylcyclohexyl diphenylphosphine,s-nmdpp,+-neomenthyldiphenylphosphine,1s,2s,5r-2-isopropyl-5-methylcyclohexyl-diphenylphosphine,1s,2s,5r-2-isopropyl-5-methylcyclohexyl diphenylphosphane,+-nmdpp,1r,3s,4s-p-menthane-3-yl diphenylphosphine,1s,2s,5r-5-methyl-2-propan-2-ylcyclohexyl-diphenylphosphane,1s,2s,5r-5-methyl-2-propan-2-yl cyclohexyl diphenyl phosphane
2734569
[(1S,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl]-diphenylphosphane
CC1CCC(C(C1)P(C2=CC=CC=C2)C3=CC=CC=C3)C(C)C
Packaging Glass bottle
Melting Point 95°C to 99°C
Color White
Quantity 500mg
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 324.25
Physical Form Crystalline Powder or Needles
Percent Purity ≥97.5%
Chemical Name or Material (S)-(+)-Neomenthyldiphenylphosphine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE