Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(-)-Nicotine, 99%

Manufacturer:  Alfa Aesar™ A1239822

Quantity 100g

 View more versions of this product

Catalog No. AAA1239822


  Check Availability
Add to cart

Specifications

Description

Solubility
Miscible with water, alcohol, ether and light petroleum.

Notes
Hygroscopic. Air and light sensitive.
Description & Specifications

Chemical Identifiers

54-11-5
162.24
UN1654
14,6524
C10H14N2
MFCD00006369
82109
(S)- (-)-Nicotine

Specifications

54-11-5
99%
162.24
UN1654
14,6524
245° to 248°C
100g
Characteristic
(S)- (-)-Nicotine
Liquid
C10H14N2
MFCD00006369
82109
10.2
−79°C
101°C (213°F)
−168° (neat)
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H310
Toxic if swallowed.
Fatal in contact with skin.

GHS P Statement
P280-P301+P310-P361-P302+P350-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NICOTINE

RTECSNumber : QS5250000

TSCA : TSCA

storageNote1 : Air Sensitive and Hygroscopic

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit