Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Nipecotic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
59045-82-8
C6H11NO2
129.16
XJLSEXAGTJCILF-YFKPBYRVSA-N
s-piperidine-3-carboxylic acid,s-+-nipecotic acid,s-nipecotic acid,3s-piperidine-3-carboxylic acid,+-nipecotic acid,s-+-3-piperidinecarboxylic acid,3s-nipecotic acid,l +-nipecotic acid,s-hexahydronicotinic acid,+-hexahydronicotinic acid
6575389
CHEBI:222169
(3S)-piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O
59045-82-8
C6H11NO2
129.16
XJLSEXAGTJCILF-YFKPBYRVSA-N
s-piperidine-3-carboxylic acid,s-+-nipecotic acid,s-nipecotic acid,3s-piperidine-3-carboxylic acid,+-nipecotic acid,s-+-3-piperidinecarboxylic acid,3s-nipecotic acid,l +-nipecotic acid,s-hexahydronicotinic acid,+-hexahydronicotinic acid
6575389
CHEBI:222169
(3S)-piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O
Melting Point 247°C to 249°C
Infrared Spectrum Authentic
Specific Rotation + 4.6
Color White
Quantity 1g
Assay Percent Range 96%
Formula Weight 129.16
Physical Form Powder
Percent Purity 96%
Chemical Name or Material (S)-(+)-Nipecotic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning