Learn More
S. Paratyphi Mouse anti-All, Unconjugated, Clone: Clone 1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma MABX874110KC
 View more versions of this product
Catalog No. MABX87110MI
Add to cart
Description
Description
S. Paratyphi Monoclonal antibody specifically detects S. Paratyphi in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.
Specifications
Specifications
S. Paratyphi
Clone 1
Affinity Purified
Mouse
IgG3
10 mg
2 to 8°C. For long-term storage, freeze at -20°C.
Monoclonal
All
ELISA, Immunodiffusion, Western Blot
Unconjugated
Anti-S. enterica paratyphi
S. paratyphi lysate
Affinity Chromatography Purified
Infectious Diseases
Primary
Specific for S. paratyphi