Learn More
S. Paratyphi Mouse anti-All, Unconjugated, Clone: Clone 1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MABX87411KC
 View more versions of this product
Catalog No. MABX87411MI
Add to Cart
Description
Description
S. Paratyphi Monoclonal antibody specifically detects S. Paratyphi in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.
Specifications
Specifications
S. Paratyphi
Monoclonal
Unconjugated
Mouse
Affinity Purified
Infectious Diseases
Specific for S. paratyphi
2 to 8°C. For long-term storage, freeze at -20°C.
ELISA, Immunodiffusion, Western Blot
Clone 1
Anti-S. enterica paratyphi
S. paratyphi lysate
1 mg
Primary
All species
IgG3
Product Suggestions
Product Suggestions