Alfa Aesar™ (S)-Phanephos, 98%, ee 98%

Manufacturer: Alfa Aesar™ H25813-MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25813MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 192463-40-4
    Molecular Formula C40H34P2
    Formula Weight 576.65
    MDL Number MFCD03094575
    Synonym (S)-(+)-4,12-Bis(diphenylphosphino)[2.2]paracyclophane
    Chemical Name or Material (S)-Phanephos

    Specifications

    Formula Weight 576.65
    CAS 192463-40-4
    Melting Point 226° to 230°C
    Quantity 250mg
    Molecular Formula C40H34P2
    MDL Number MFCD03094575
    Synonym (S)-(+)-4,12-Bis(diphenylphosphino)[2.2]paracyclophane
    Chemical Name or Material (S)-Phanephos
    Assay 98%, ee 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit