Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Prolinol, 98%, Thermo Scientific Chemicals

Catalog Number p-7045199
Click to view available options
Quantity:
1 g
5 g
25 g
23356-96-9
C5H11NO
101.149
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
640091
[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)CO
23356-96-9
C5H11NO
101.149
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
640091
[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)CO

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water. Soluble in chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Refractive Index 1.4853
Density 1.025
Boiling Point 74°C to 76°C (2mmHg)
Melting Point 42°C to 44°C
Quantity 1 g
Flash Point 86°C (186°F)
Optical Rotation +31° (c=1 in Toluene)
Sensitivity Air sensitive; Hygroscopic
Beilstein 79843
Solubility Information Fully miscible in water. Soluble in chloroform.
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material (S)-(+)-Prolinol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 245-605-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit