Alfa Aesar™ (S)-(-)-Propylene oxide, 99%

Manufacturer: Alfa Aesar™ B2234122

Quantity 100g

 View more versions of this product

Catalog No. AAB2234122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16088-62-3
    Molecular Formula C3H6O
    Formula Weight 58.08
    MDL Number MFCD00064312
    UN Number UN1280
    Beilstein 79765
    Merck Index 14,7856
    Synonym (S)-1,2-Epoxypropane; (S)-Methyloxirane
    Chemical Name or Material (S)-(-)-Propylene oxide

    Specifications

    Formula Weight 58.08
    MDL Number MFCD00064312
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number UJ2651000
    Molecular Formula C3H6O
    CAS 16088-62-3
    Merck Index 14,7856
    Boiling Point 34° to 35°C
    Quantity 100g
    Flash Point −37°C (−35°F)
    Odor Ether-like
    Optical Rotation −14° (Neat)
    DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: PROPYLENE OXIDE
    UN Number UN1280
    Beilstein 79765
    Synonym (S)-1,2-Epoxypropane; (S)-Methyloxirane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H224-H340-H350-H302-H312-H332-H315-H319-H335
    Extremely flammable liquid and vapour.
    May cause genetic defects.
    May cause cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material (S)-(-)-Propylene oxide
    Assay 99%

    Description

    Applications
    (S)-(-)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used in making the polyurethane plastics. It finds an application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It is also used in model aircraft and surface vehicles as a glow fuel.

    Solubility
    Miscible with water and many organic solvents.

    Notes
    Highly flammable. Keep away from sources of ignition. The substance may polymerize under the influence of bases, acids and metal chlorides.

    GHS H Statement
    H224-H340-H350-H302-H312-H332-H315-H319-H335
    Extremely flammable liquid and vapour.
    May cause genetic defects.
    May cause cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit