Alfa Aesar™ (S,S)-DIPAMP, 97%

Manufacturer: Alfa Aesar™ H28037MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28037MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97858-62-3
    Molecular Formula C28H28O2P2
    Formula Weight 458.47
    MDL Number MFCD05863547
    Synonym (1S,2S)-(+)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
    Chemical Name or Material (S,S)-DIPAMP

    Specifications

    Formula Weight 458.47
    Molecular Formula C28H28O2P2
    MDL Number MFCD05863547
    Synonym (1S,2S)-(+)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
    Melting Point 101° to 105°C
    Quantity 250mg
    Optical Rotation +84° (c=1 in Chloroform)
    CAS 97858-62-3
    Chemical Name or Material (S,S)-DIPAMP
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit