Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-(-)-Hydrobenzoin, 98+%, Thermo Scientific Chemicals

Catalog Number AAL1408206
Click to view available options
Quantity:
5 g
25 g
2325-10-2
MFCD00064255
2325-10-2
MFCD00064255

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 148°C to 150°C
Color White
Molecular Formula C11H16ClNO2
Quantity 5 g
Beilstein 2330888
Merck Index 14,4777
Synonym 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N
SMILES COC1=C(C=C2CNCCC2=C1)OC.Cl
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
Molecular Weight (g/mol) 229.70
PubChem CID 6456035
Formula Weight 214.27
Percent Purity ≥98%
Physical Form Crystals
Chemical Name or Material (S,S)-(-)-Hydrobenzoin
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit