Alfa Aesar™ [(S,S)-TsDPEN-Ru(mesitylene)Cl]

Manufacturer: Alfa Aesar™ H37991-03

Quantity 1g

 View more versions of this product

Catalog No. AAH3799103

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 174813-81-1
    Formula Weight 622.18
    MDL Number MFCD03791032
    Chemical Name or Material [(S,S)-TsDPEN-Ru(mesitylene)Cl]

    Specifications

    Formula Weight 622.18
    Physical Form Powder
    MDL Number MFCD03791032
    CAS 174813-81-1
    Color Brown
    Quantity 1g
    Chemical Name or Material [(S,S)-TsDPEN-Ru(mesitylene)Cl]

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit