Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ S·Tag Thrombin Purification Kit

Catalog No. 692323
Click to view available options
Quantity:
1 Kit

Application in protein purification

  • 2mL S-protein Agarose
  • 3 × 5mL 10X Bind/Wash Buffer
  • 3 × 1mL 10X Thrombin Cleavage Buffer
  • 50U Biotinylated Thrombin
  • 0.4mL Streptavidin Agarose
  • 2ea/pkg Spin Filters, 5mL capacity
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Multiple storage requirements
Quantity 1 Kit
Product Type Thrombin Purification Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.