Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-tert-Butylsulfinamide, 97+%, (>98.0% ee), ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
343338-28-3
C4H11NOS
121.2
MFCD05861480
CESUXLKAADQNTB-ZETCQYMHSA-N
s---2-methyl-2-propanesulfinamide,s-2-methylpropane-2-sulfinamide,s-tert-butanesulfinamide,s---tert-butanesulfinamide,s---t-butylsulfinamide,s---tert-butylsulfinamide,s-2-methyl-2-propanesulfinamide,s-tert-butylsulfinamide,s---t-butylmethylsulfinamide,s-t-butyl sulfinamide
11355477
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
343338-28-3
C4H11NOS
121.2
MFCD05861480
CESUXLKAADQNTB-ZETCQYMHSA-N
s---2-methyl-2-propanesulfinamide,s-2-methylpropane-2-sulfinamide,s-tert-butanesulfinamide,s---tert-butanesulfinamide,s---t-butylsulfinamide,s---tert-butylsulfinamide,s-2-methyl-2-propanesulfinamide,s-tert-butylsulfinamide,s---t-butylmethylsulfinamide,s-t-butyl sulfinamide
11355477
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
Melting Point 101°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Enantiomeric Excess 98.0% min.
Color White
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula (CH3)3CSONH2
Merck Index 15, 3607
Formula Weight 121.2
Physical Form Crystalline Powder
Percent Purity ≥97.0%
Chemical Name or Material (S)-tert-Butylsulfinamide
Show More Show Less