Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-tert-Leucinol, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
112245-13-3
C6H15NO
117.192
MFCD00192250
JBULSURVMXPBNA-RXMQYKEDSA-N
l-tert-leucinol,s-tert-leucinol,2s-2-amino-3,3-dimethylbutan-1-ol,2s-2-amino-3,3-dimethyl-butan-1-ol,s-2-amino-3,3-dimethylbutanol,s-2-amino-3,3-dimethyl-1-hydroxybutane,r---tert-leucinol,1-butanol, 2-amino-3,3-dimethyl-, 2s
2734079
(2S)-2-amino-3,3-dimethylbutan-1-ol
CC(C)(C)C(CO)N
112245-13-3
C6H15NO
117.192
MFCD00192250
JBULSURVMXPBNA-RXMQYKEDSA-N
l-tert-leucinol,s-tert-leucinol,2s-2-amino-3,3-dimethylbutan-1-ol,2s-2-amino-3,3-dimethyl-butan-1-ol,s-2-amino-3,3-dimethylbutanol,s-2-amino-3,3-dimethyl-1-hydroxybutane,r---tert-leucinol,1-butanol, 2-amino-3,3-dimethyl-, 2s
2734079
(2S)-2-amino-3,3-dimethylbutan-1-ol
CC(C)(C)C(CO)N
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Specific Rotation +34° to +40° (25°C, 589nm) (c=1.5, EtOH)
Density 90g/mL
Specific Rotation Condition + 37.50 (20.00°C c=2,EtOH)
Quantity 5g
Color White or Colorless
Assay Percent Range 95%
Linear Formula (CH3)3CCH(NH2)CH2OH
Formula Weight 117.19
Physical Form Crystalline Low Melting Mass or Liquid
Percent Purity ≥94%
Chemical Name or Material (S)-tert-Leucinol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.