Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(-)-Tetrahydro-3-furoic acid, 97%

Click to view available options
Quantity:
250mg
1g
168395-26-4
C5H8O3
116.116
MFCD07369984
BOTREHHXSQGWTR-BYPYZUCNSA-N
s-tetrahydrofuran-3-carboxylic acid,3s-oxolane-3-carboxylic acid,s-tetrahydro-3-furancarboxylic acid,s-tetrahydro-3-furoic acid,s-tetrahydro-furan-3-carboxylic acid,3s-tetrahydrofuran-3-carboxylic acid,l-tetrahydrofuran-3-carboxylic acid,3-furancarboxylic acid, tetrahydro-, 3s,3s-3-oxolanecarboxylic acid,s, ?-tetrahydro-3-furancarboxylic acid
40784874
(3S)-oxolane-3-carboxylic acid
C1COCC1C(=O)O
168395-26-4
C5H8O3
116.116
MFCD07369984
BOTREHHXSQGWTR-BYPYZUCNSA-N
s-tetrahydrofuran-3-carboxylic acid,3s-oxolane-3-carboxylic acid,s-tetrahydro-3-furancarboxylic acid,s-tetrahydro-3-furoic acid,s-tetrahydro-furan-3-carboxylic acid,3s-tetrahydrofuran-3-carboxylic acid,l-tetrahydrofuran-3-carboxylic acid,3-furancarboxylic acid, tetrahydro-, 3s,3s-3-oxolanecarboxylic acid,s, ?-tetrahydro-3-furancarboxylic acid
40784874
(3S)-oxolane-3-carboxylic acid
C1COCC1C(=O)O
Color Yellow to Brown
Quantity 1g
Beilstein 1281203
Formula Weight 116.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (S)-(-)-Tetrahydro-3-furoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning