Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-Timolol maleate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
26921-17-5
C13H25N4O3S+
317.428
BLJRIMJGRPQVNF-JTQLQIEISA-O
tert-butyl-2s-2-hydroxy-3-4-morpholin-4-yl-1,2,5-thiadiazol-3-yl oxy propyl azanium,tert-butyl-2s-2-hydroxy-3-4-4-morpholinyl-1,2,5-thiadiazol-3-yl oxy propyl ammonium,tert-butyl-2s-3-4-morpholin-4-yl-1,2,5-thiadiazol-3-yl oxy-2-oxidanyl-propyl azanium
6919026
tert-butyl-[(2S)-2-hydroxy-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propyl]azanium
CC(C)(C)[NH2+]CC(COC1=NSN=C1N2CCOCC2)O
This item is not returnable. View return policy
26921-17-5
C13H25N4O3S+
317.428
BLJRIMJGRPQVNF-JTQLQIEISA-O
tert-butyl-2s-2-hydroxy-3-4-morpholin-4-yl-1,2,5-thiadiazol-3-yl oxy propyl azanium,tert-butyl-2s-2-hydroxy-3-4-4-morpholinyl-1,2,5-thiadiazol-3-yl oxy propyl ammonium,tert-butyl-2s-3-4-morpholin-4-yl-1,2,5-thiadiazol-3-yl oxy-2-oxidanyl-propyl azanium
6919026
tert-butyl-[(2S)-2-hydroxy-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propyl]azanium
CC(C)(C)[NH2+]CC(COC1=NSN=C1N2CCOCC2)O

β1 antagonist

β1-adrenergic blocker.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 432.49
Percent Purity >98%
Chemical Name or Material (S)-Timolol maleate

Recommended Storage : Store at Room Temperature