Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

s-Trioxane, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2kg
5kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
110-88-3
C3H6O3
90.08
MFCD00006563
BGJSXRVXTHVRSN-UHFFFAOYSA-N
s-trioxane,trioxymethylene,metaformaldehyde,triformol,formaldehyde, trimer,aldeform,formagene,marvosan,trioxan,sym-trioxane
8081
CHEBI:38043
1,3,5-trioxane
C1OCOCO1
110-88-3
C3H6O3
90.08
MFCD00006563
BGJSXRVXTHVRSN-UHFFFAOYSA-N
s-trioxane,trioxymethylene,metaformaldehyde,triformol,formaldehyde, trimer,aldeform,formagene,marvosan,trioxan,sym-trioxane
8081
CHEBI:38043
1,3,5-trioxane
C1OCOCO1
Specific Gravity 1.17
Color Colorless to White
Sublimation Point 0115.0°C
Quantity 100g
Flash Point 45°C
Infrared Spectrum Authentic
Melting Point 61.0°C to 62.0°C
Density 1.1700g/mL
Packaging Plastic bottle
Boiling Point 114.0°C to 116.0°C
Assay Percent Range 99+%
Beilstein 19,381
Merck Index 15,9906
Solubility Information Solubility in water: soluble in water: 221g/L (25°C). Other solubilities: soluble in alcohols,ether,ketones,acetone,,chlorinatid and aromatic hydrocarbons,and,other organic solvents,slightly soluble in pentane and petr.ether
Formula Weight 90.08
Physical Form Crystals or Crystalline Flakes
Percent Purity ≥99%
Chemical Name or Material s-Trioxane
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Suspected of damaging the unborn child.
Flammable solid.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger