Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ HU-210 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco S0241ML
Description
HU-210 is a synthetic cannabinoid found in the herbal mixture 'Spice'. Illicit use of this Spice cannabinoid includes smoking the herbal product to obtain a psychoactive effect. Structurally similar to (-)-δ8-THC and 11-Hydroxy-δ9-THC, HU-210 is considerably more potent than natural THC from cannabis and has an extended duration of action. In July, 2009, the US Drug Enforcement Agency (DEA) placed HU-210 on its Drugs and Chemicals of Concern list. Since HU-210's placement on this list, DEA has reported seizures of Spice herbal mixtures in Ohio and Florida. HU-210 is currently listed as a Schedule I controlled substance. In recognition of this regulatory challenge, Cerilliant supplies its certified HU-210 reference standard in a convenient, quantitative, DEA-exempt solution.Specifications
67-56-1, 112830-95-2 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
386.57 | |
1 mL |
C25H38O3 | |
100 μg/mL in methanol | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction