Learn More
Thermo Scientific™ S1 Pipet Filler Accessories
GREENER_CHOICE
Catalog No. p-4530111
Click to view available options
Type:
Nosepiece
Pipette Filter, Non-sterile
Pipette Filter, Sterile
Silicone Pipet Gripper
Table Mount Stand
Universal Power Supply
Wall Mount Stand
Wing Stand
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
TRUSTED_SUSTAINABILITY
Specifications
Description Hydrophobic Filters, 0.45μm, Non-Sterile
For Use With S1 Pipette Filler
Type Pipette Filter, Non-sterile
Includes Includes 5 pieces of filters
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.