Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100 calcium binding protein A14 Antibody (OTI2G9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273966G
Description
S100 calcium binding protein A14 Monoclonal specifically detects S100 calcium binding protein A14 in Human, Mouse samples. It is validated for Western Blot.Specifications
S100 calcium binding protein A14 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
S100A14 | |
Full length recombinant protein of human S100 calcium binding protein A14 (NP_065723) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot | |
OTI2G9 | |
Western Blot | |
protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114 | |
Mouse | |
Affinity Purified | |
Cancer | |
57402.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction