Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ S100 Protein Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ S100 Protein Ab-1, Mouse Monoclonal Antibody..

$204.00 - $603.50

Specifications

Antigen S100 Protein Ab-1
Clone 4C4.9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
View More Specs

Products 11
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS296B
View Documents
Thermo Scientific™
MS296B
Biotinylated Ab with BSA and Azide 1mL
Each for $603.50
Add to cart
 
MS296B0
View Documents
Thermo Scientific™
MS296B0
Biotinylated Ab with BSA and Azide 100μL
Each for $204.00
Add to cart
 
MS296B1
View Documents
Thermo Scientific™
MS296B1
Biotinylated Ab with BSA and Azide 500μL
Each for $544.50
Add to cart
 
MS296P
View Documents
Thermo Scientific™
MS296P
Purified Ab with BSA and Azide 1mL
Each for $505.50
Add to cart
 
MS296P1
View Documents
Thermo Scientific™
MS296P1
Purified Ab with BSA and Azide 500μL
Each for $357.00
Add to cart
 
MS296P1ABX
View Documents
Thermo Scientific™
MS296P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $363.00
Add to cart
 
MS296PABX
View Documents
Thermo Scientific™
MS296PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $444.00
Add to cart
 
MS296RQ
View Documents
Thermo Scientific™
MS296RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $412.50
Add to cart
 
MS296R1
View Documents
Thermo Scientific™
MS296R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS296R7
View Documents
Thermo Scientific™
MS296R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS296P0
View Documents
Thermo Scientific™
MS296P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ S100 Protein Ab-1, Mouse Monoclonal Antibody.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Mouse

Clone: 4C4.9

Isotype: IgG2a

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified bovine brain S100 protein

Molecular Weight: ~22kDa (nonreduced); ~11kDa (reduced)

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

S100 Protein Ab-1
Immunohistochemistry (Paraffin)
Murine
Neuroscience
Purified bovine brain S100 protein
4C4.9
Monoclonal
IVD
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit