Learn More
Epredia™ Lab Vision™ S100 Protein Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Epredia™ S100 Protein Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results.
Specifications
Antigen | S100 Protein Ab-2 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB044A1
|
Epredia™
RB044A1 |
Concentrated | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
RB044R1
|
Epredia™
RB044R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
RB044R7
|
Epredia™
RB044R7 |
Prediluted | 7 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Cow, Kangaroo, Mouse, Rat and Chicken. Others not tested.
Epitope: Not determined
Immunogen: S100 protein purified from cow brain
Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)
Positive Control: Melanoma or Schwannoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunocytochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
S100 Protein Ab-2 | |
Polyclonal | |
Rabbit | |
Cancer and Tumor Biology | |
S100 protein purified from cow brain | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Bovine, Chicken, Human, Kangaroo, Mouse | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.