Learn More
S100 ProteinRabbit anti-Bovine, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 200401874

Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against bovine S100 protein. A BLAST analysis was used to suggest reactivity with this protein from bovine based on 100% homology with the immunogen sequence. A 98% homology is noted for S100 alpha chain from primate sources. Mouse, rat and dog show 94% homology with the bovine S100 alpha sequence. Expect cross reactivity with S100 from most mammalian sources. Cross reactivity with S100 from other specific sources has not been determined. Homologies for the S100 beta chain are similar.
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Specifications
S100 Protein | |
Polyclonal | |
Unconjugated | |
S100B | |
AI266795; AI850290; BOS_1447; BOS_2809; Bpb; NEF; Protein S100-A1; Protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100A; S100a1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
Rabbit | |
500 μg | |
Primary | |
Bovine | |
Antibody |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
5 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
P02638, P02639 | |
S100A1, S100B | |
This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full-length bovine S100 protein (mixture of aa homodimers and ab heterodimers). | |
RUO | |
525716, 528735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.