Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A13 Rabbit anti-Mouse, Clone: 184, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289547100UL
Description
S100A13 Monoclonal specifically detects S100A13 in Mouse samples. It is validated for ELISA.Specifications
S100A13 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
6284 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA | |
184 | |
ELISA 1:5000-1:10000 | |
S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
Recombinant Mouse S100A13 Protein | |
100 μg | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction