Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550783
Description
The antibody detects endogenous levels of total S100A14 protein.
S100A14 Polyclonal antibody specifically detects S100A14 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
S100A14 | |
Polyclonal | |
Unconjugated | |
S100a14 | |
1110013O05Rik; BCMP84; breast cancer membrane protein 84; Gm1020; Protein S100-A14; S100 calcium binding protein A14; S100 calcium binding protein A15; S100 calcium-binding protein A14; S100A14; S100a15; S114 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57402 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9HCY8 | |
S100a14 | |
Full length fusion protein of Human S100A14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction