Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100A16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580987
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human S100A16.
S100A16 (S100 Calcium Binding Protein A16) is a Protein Coding gene. Diseases associated with S100A16 include Robinow Syndrome, Autosomal Recessive and Robinow Syndrome. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity. Calcium-binding protein. Binds one calcium ion per monomer. Can promote differentiation of adipocytes (in vitro). Overexpression in preadipocytes increases their proliferation, enhances adipogenesis and reduces insulin-stimulated glucose uptake.
Specifications
S100A16 | |
Polyclonal | |
Unconjugated | |
S100a16 | |
2300002L21Rik; AAG13; aging-associated gene 13 protein; aging-associated protein 13; AI325039; AI663996; DT1P1A7; MGC17528; protein S100-A16; Protein S100F; Protein S100-F; S100 calcium binding protein A16; S100 calcium-binding protein A16; S100A16; S100F | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
140576 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
5 mg/mL | |
PBS with no preservative | |
Q96FQ6 | |
S100a16 | |
Recombinant Human S100A16 protein (Met1-Ser103). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction