Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A16 Rabbit anti-Human, Clone: 140, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | S100A16 |
---|---|
Clone | 140 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
S100A16 Monoclonal specifically detects S100A16 in Human samples. It is validated for ELISA.Specifications
S100A16 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
140576 | |
IgG | |
Protein A purified |
140 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
Aging-associated gene 13 protein, aging-associated protein 13, DT1P1A7, MGC17528, protein S100-A16, Protein S100-F, S100 calcium binding protein A16, S100FS100 calcium-binding protein A16 | |
Recombinant Human S100A16 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title