Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A2, Mouse anti-Human, Clone: CPTC-S100A2-2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279764100UG
Description
S100A2 Monoclonal specifically detects S100A2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
S100A2 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P29034, P29034 | |
S100A2 | |
Recombinant full-length human S100A2 protein | |
100 μg | |
Cancer, Cell Biology, Cellular Markers, Neuroscience | |
6273 | |
Store at 4°C. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CPTC-S100A2-2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
CAN19protein S100-A2, MGC111539, S100 calcium binding protein A2, S100 calcium-binding protein A2Protein S-100L, S100L | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction