Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A4 Mouse anti-Human, Clone: S100A4/1481, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14977

Catalog No. 89-034-997


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human S100A4.

  • High specificity
  • High reactivity
Specifications

Specifications

S100A4
S100A4/1481
Mouse monoclonal antibody raised against full length recombinant human S100A4.
In 10 mM PBS.
Liquid
S100A4
Recombinant protein corresponding to full length human S100A4.
Protein A/G purification
RUO
Primary
6275
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determine
P26447
18A2|42A|CAPL|FSP1|MTS1|P9KA|PEL98
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)