Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A4 Mouse anti-Human, Mouse, Allophycocyanin/Cy7, Clone: S100A4/1481, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254580APCCY7

Catalog No. N254580APC7


Add to cart

Description

Description

S100A4 Monoclonal antibody specifically detects S100A4 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

S100A4
S100A4/1481
Flow Cytometry
Conjugated, Purified
S100A4
Recombinant human S100A4 protein
Protein A or G purified
Cancer
Primary
6275.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
18A2, Calvasculin, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog)
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit