Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A5 Rabbit anti-Mouse, DyLight 755, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298277IR
Description
S100A5 Polyclonal specifically detects S100A5 in Mouse samples. It is validated for ELISA.Specifications
S100A5 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
6276 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 755 | |
Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
Produced in rabbits immunized with purified, recombinant S100A5 (P63084; Met 1-Lys 93). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title