Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A8/A9 Antibody (5.5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250283APC
Description
S100A8/A9 Monoclonal specifically detects S100A8/A9 in Human samples. It is validated for Flow Cytometry.Specifications
S100A8/A9 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
S100A8 | |
Acute monocytic leukaemia cells. | |
0.1 mL | |
Cancer | |
6279 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
5.5 | |
Flow Cytometry | |
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8, S100 calcium binding protein A8, S100A8 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction