Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Mouse anti-Human, DyLight 405, Clone: 7C12/4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151517V

Catalog No. NBP151517V


Add to cart

Description

Description

S100A8 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Flow Cytometry, Western Blotting.
Specifications

Specifications

S100A8
7C12/4
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
S100A8
Recombinant human S100A8
IgM purified
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Mouse
IgM
0.1 ml
Cancer
Primary
6279
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit