Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Mouse anti-Human, DyLight™ 650, Clone: 2C5/4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP276650C

Catalog No. NBP276650C


Add to cart

Description

Description

S100A8 Monoclonal antibody specifically detects S100A8 in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

S100A8
2C5/4
Western Blot, ELISA
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody is specific for S100A8 both as an isolated protein and in combination with S100A9.
ELISA, Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
S100A8
Recombinant human S100A8
0.1 mL
Primary
6279
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit