Learn More
Abnova™ S100a8 (Mouse) ELISA Kit
Use to measure quantitatively mouse S100a8. Sandwich enzyme immunoassay.
Manufacturer:  Abnova™ KA5087
Catalog No. 89-334-071
Add to cart
Description
Description
S100a8 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of mouse S100a8 in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).
Specifications
Specifications
ELISA Kit
20201
1 Kit
Quant
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Mouse