Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Rabbit anti-Human, DyLight 755, Clone: 203, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289806IR

Catalog No. NB109378


Add to cart

Description

Description

S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Western Blot.
Specifications

Specifications

S100A8
203
50 mM Sodium Borate
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A8 (NP_002955.2; Met1-Glu93).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 755
Purified
Rabbit
IgG
0.1 mL
Cancer
Primary
6279
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit