Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Rat anti-Mouse, Alexa Fluor™ 350, Clone: 63N13G5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227067AF350

Catalog No. NP227067A30


Add to cart

Description

Description

S100A8 Monoclonal antibody specifically detects S100A8 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

S100A8
63N13G5
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
S100A8
Within the range of amino acids 2-98 of mouse S100A8 were used as the immunogen for the antibody.
Protein G purified
Cancer
Primary
6279.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Rat
IgG2b Lambda
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit