Learn More
S100A8 Rat anti-Mouse, Alexa Fluor™ 532, Clone: 63N13G5, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227067AF532
Catalog No. NP227067A32
Add to cart
Description
Description
S100A8 Monoclonal antibody specifically detects S100A8 in Mouse samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
S100A8
63N13G5
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Rat
IgG2b Lambda
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
S100A8
Within the range of amino acids 2-98 of mouse S100A8 were used as the immunogen for the antibody.
0.1 mL
Primary
6279