Sino Biological™ S100A8 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human S100A8 (Leu 2-Glu 93) fused to polyhistidine tag at N-terminus

Manufacturer: Sino Biological™ 11138H08B50

 View more versions of this product

Catalog No. 11-138-H08B50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 12.2kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Human
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name S100 Calcium Binding Protein A8
    Gene Symbol S100A8
    Protein Tag His Tag, MBP (Maltose Binding Protein)
    Validated Application Assay Standard
    Protein Family Other proteins
    Expression System Insect
    Product Type S100A8 Recombinant Human Protein
    Format Lyophilized

    Expressed from DNA sequence encoding human S100A8 (Leu 2-Glu 93) fused to polyhistidine tag at N-terminus

    • Measured by its ability to bind recombinant human S100A9 in functional ELISA
    • Measured by its ability to inhibit t proliferation of MCF7 human breast adenocarcinoma cells
    • ED50 for effect is 3-15µg/mL
    • Lyophilized in 140mM

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit