Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ S100A8 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human S100A8 (Leu 2-Glu 93) fused to polyhistidine tag at N-terminus

Manufacturer:  Sino Biological™ 11138H08B5

 View more versions of this product

Catalog No. 11-138-H08B5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human S100A8 (Leu 2-Glu 93) fused to polyhistidine tag at N-terminus

  • Measured by its ability to bind recombinant human S100A9 in functional ELISA
  • Measured by its ability to inhibit t proliferation of MCF7 human breast adenocarcinoma cells
  • ED50 for effect is 3-15µg/mL
  • Lyophilized in 140mM
Description & Specifications

Specifications

Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
S100A8
Other proteins
S100A8 Recombinant Human Protein
12.2kd
5μg
Recombinant
S100 Calcium Binding Protein A8
Assay Standard
Insect
His Tag, MBP
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit