Learn More
S100A8 Recombinant Protein, Abnova Available on GSA/VA Contract for Federal Government customers only.
Human S100A8 full-length ORF recombinant protein with GST-tag at N-terminal
Manufacturer:  Abnova Corporation H00006279P0110
 View more versions of this product
Catalog No. 89-010-631
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human S100A8 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
35.97
8
Glutathione Sepharose 4 Fast Flow
10μg
-80°C
Yes