Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Mouse anti-Human, Mouse, Rat, Bovine, Allophycocyanin/Cy7, Clone: S100B/1012, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259619APCCY7

Catalog No. N259619APC7


Add to cart

Description

Description

S100B Monoclonal antibody specifically detects S100B in Human, Mouse, Rat, Bovine samples. It is validated for Flow Cytometry.
Specifications

Specifications

S100B
S100B/1012
Flow Cytometry
Conjugated, Purified
S100B
Recombinant full-length human S100B protein
Protein A or G purified
Apoptosis, Biologically Active Proteins, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells
Primary
6285.0
Bovine, Human, Mouse, Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
The specificity of this monoclonal antibody to S100B was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and a beta chain whereas S100B is composed of two beta chains. This antibody is specific against an epitope located on the beta -chain (i.e. in S-100A and S-100B) but not on the alpha-chain of S-100 (i.e. in S-100A and S100A0). This antibody can be used to localize S-100A and S-100B in various tissue sections. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lip ocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells. Neoplasms derived from these cells also express S-100 protein, albeit non-uniformly. A large number of well-differentiated tumors of the salivary gland, adipose and cartilaginous tissue, and Schwann cell -derived tumors express S-100 protein. Almost all malignant melanomas and cases of histiocytosis X are positive for S -100 protein
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit