Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Mouse anti-Human, Mouse, Rat, Clone: SA-12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141373SS

 View more versions of this product

Catalog No. NBP141373SS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

S100B Monoclonal antibody specifically detects Antigen in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

S100B
SA-12
Unconjugated
Lyophilized from 1.2% sodium acetate, with 2mg BSA and 0.01mg NaN3 as preservative. with 0.01 mg Sodium Azide
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Mouse
IgG1
0.025 mg
Apoptosis, Cytoskeleton Markers, Neuroscience, Stem Cells
Primary
6285
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1.0 mg/mL
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunocytochemistry
Purified
S100B
Bovine brain S-100b.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute 0.1mg size with 0.1ml of deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit