Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Rabbit anti-Human, Mouse, Rat, Bovine, PE-Cyanine7, Clone: S100B/1706R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254426PECY7

Catalog No. N254426PEC7


Add to cart

Description

Description

S100B Monoclonal antibody specifically detects S100B in Human, Mouse, Rat, Bovine samples. It is validated for Flow Cytometry.
Specifications

Specifications

S100B
S100B/1706R
Flow Cytometry
Conjugated, Purified
S100B
Recombinant full-length human S100B protein
Protein A purified
Apoptosis, Biologically Active Proteins, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells
Primary
6285.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Bovine, Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit