Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100P Rabbit anti-Human, Clone: S100P/4386R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30774610UG

 View more versions of this product

Catalog No. NB122965


Add to cart

Description

Description

S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

S100P
S100P/4386R
Unconjugated
10 mM PBS with 0.05% BSA
MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E
Recombinant fragment corresponding to C-terminal of human S100P protein
Protein A or G purified
RUO
Primary
6286
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Rabbit
IgG
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit